Learn More
Macherey-Nagel™ Optima™ 5 MS GC Capillary Column
Product Code. 10479161
Click to view available options
Length (Metric):
12 m
15 m
25 m
30 m
50 m
60 m
Diameter (Metric) Inner:
0.2 mm
0.25 mm
0.32 mm
Film Thickness:
0.2 μm
0.25 μm
0.35 μm
0.5 μm
1 μm
Unit Size:
Each
Product Code. 10479161
Brand: Macherey-Nagel™ 726220.15
Only null left
Specifications
Diameter (Metric) 0.4 mm
Type GC Capillary Column
Diameter (Metric) Inner 0.25 mm
Stationary Phase Optima™ 5 MS
USP Type G27, G36
For Use With (Application) GC/MS, ECD
Temperature Range 320°C to 340°C
Film Thickness 0.25 μm
Length (Metric) 15 m
Quantity 1 Each