Learn More
Macherey-Nagel™ Optima™ 5 High Performance GC Capillary Column
Click to view available options
Length (Metric):
10 m
60 m
Diameter (Metric) Inner:
0.1 mm
0.25 mm
missing translation for 'unitSize'
Each
Specifications
Specifications
| Diameter (Metric) | 0.4 mm |
| Type | High Performance GC Capillary Column |
| Diameter (Metric) Inner | 0.1 mm |
| Stationary Phase | Optima™ 5 |
| USP Type | G27, G36 |
| For Use With (Application) | Gas Chromatography (GC) |
| Temperature Range | 340°C to 360°C |
| Film Thickness | 0.1 μm |
| Length (Metric) | 10 m |
| Quantity | 1 Each |