Learn More
Macherey-Nagel™ Optima™ 5 High Performance GC Capillary Column
Product Code. 10461472
Click to view available options
Length (Metric):
10 m
15 m
25 m
30 m
50 m
60 m
Diameter (Metric) Inner:
0.2 mm
0.25 mm
0.53 mm
Film Thickness:
0.1 μm
0.2 μm
0.25 μm
0.35 μm
0.5 μm
1 μm
2 μm
5 μm
Unit Size:
Each
Pack of 10
Product Code. 10461472
Brand: Macherey-Nagel™ 726916.30
Only null left
Specifications
Diameter (Metric) 0.8 mm
Type High Performance GC Capillary Column
Diameter (Metric) Inner 0.53 mm
Stationary Phase Optima™ 5
USP Type G27
For Use With (Application) Gas Chromatography (GC)
Temperature Range 340°C
Film Thickness 5 μm
Length (Metric) 30 m
Quantity 1 Each