Learn More
Macherey-Nagel™ Optima™ 240 GC Capillary Column
Click to view available options
Length (Metric):
25 m
30 m
50 m
60 m
Diameter (Metric) Inner:
0.25 mm
0.32 mm
Film Thickness:
0.25 μm
0.35 μm
0.5 μm
Unit Size:
Each
Specifications
Specifications
| Diameter (Metric) | 0.4 mm |
| Type | GC Capillary Column |
| Diameter (Metric) Inner | 0.25 mm |
| Stationary Phase | Optima™ 240 |
| For Use With (Application) | Gas Chromatography (GC), Dioxins |
| Temperature Range | 260°C to 280°C |
| Film Thickness | 0.5 μm |
| Length (Metric) | 60 m |
| Quantity | 1 Each |