Learn More
Macherey-Nagel™ Optima™ 240 GC Capillary Column
Product Code. 10452042
Click to view available options
Length (Metric):
25 m
30 m
50 m
60 m
Diameter (Metric) Inner:
0.25 mm
0.32 mm
Film Thickness:
0.25 μm
0.35 μm
0.5 μm
Unit Size:
Each
Product Code. 10452042
Brand: Macherey-Nagel™ 726090.30
Only null left
Specifications
Diameter (Metric) 0.4 mm
Type GC Capillary Column
Diameter (Metric) Inner 0.25 mm
Stationary Phase Optima™ 240
For Use With (Application) Gas Chromatography (GC), Dioxins
Temperature Range 260°C to 280°C
Film Thickness 0.5 μm
Length (Metric) 30 m
Quantity 1 Each