Learn More
Macherey-Nagel™ Optima™ 1701 High Performance GC Capillary Column
Product Code. 10526681
Click to view available options
Length (Metric):
10 m
15 m
25 m
30 m
50 m
60 m
Diameter (Metric) Inner:
0.2 mm
0.25 mm
0.32 mm
0.53 mm
Film Thickness:
0.25 μm
0.35 μm
0.5 μm
1 μm
Unit Size:
Each
Pack of 50
Product Code. 10526681
Brand: Macherey-Nagel™ 726058.15
Only null left
Specifications
Diameter (Metric) 0.4 mm
Type High Performance GC Capillary Column
Diameter (Metric) Inner 0.25 mm
Stationary Phase Optima™ 1701
USP Type G46
For Use With (Application) Gas Chromatography (GC)
Temperature Range 300°C to 320°C
Film Thickness 0.25 μm
Length (Metric) 15 m
Quantity 1 Each