Learn More
Macherey-Nagel™ Optima™ 1 MS Accent GC Capillary Column
Product Code. 12462810
Click to view available options
Length (Metric):
15 m
25 m
30 m
50 m
60 m
Diameter (Metric) Inner:
0.2 mm
0.25 mm
0.32 mm
Film Thickness:
0.2 μm
0.25 μm
0.5 μm
Unit Size:
Each
Product Code. 12462810
Brand: Macherey-Nagel™ 725802.30
Specifications
Diameter (Metric) 0.5 mm
For Use With (Equipment) Ideal for Ion Trap and Quadrupole MS Detectors
Type GC Capillary Column
Diameter (Metric) Inner 0.32 mm
Stationary Phase Optima™ 1 MS Accent
USP Type G1, G2, G38
For Use With (Application) Gas Chromatography (GC)
Temperature Range 360°C
Film Thickness 0.25 μm
Length (Metric) 30 m
Quantity 1 Each