Learn More
Thermo Scientific™ Opti-Link™ Carboxylate-Modified Particles
Product Code. p-4530466 Shop All Thermo Scientific Products
Change view
Click to view available options
Diameter (Metric):
0.2 μm
0.3 μm
0.4 μm
0.5 μm
0.85 μm
0.9 μm
2 μm
3 μm
4.0 μm
5.0 μm
Quantity:
15 mL
100 mL
1000 mL
Surface Acid Loading:
High Acid
Low Acid
Medium Acid
Concentration:
10%
4%
Unit Size:
1000mL
100mL
15mL
Each
30 product options available for selection
Product selection table with 30 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Quantity Surface Acid Loading Concentration unitSize
11819922 2 μm 15 mL High Acid 10% 15mL
11829922 2 μm 100 mL High Acid 10% 100mL
11950085 3 μm 15 mL High Acid 10% 15mL
11960085 3 μm 100 mL High Acid 10% 100mL
12406462 0.2 μm 15 mL High Acid 10% 15mL
12416462 0.2 μm 100 mL High Acid 10% 100mL
12612550 0.2 μm 15 mL Medium Acid 10% 15mL
11976558 0.2 μm 100 mL Medium Acid 10% Each
12446462 0.2 μm 100 mL Medium Acid 10% 100mL
11894153 0.3 μm 15 mL High Acid 10% 15mL
11804163 0.3 μm 100 mL High Acid 10% 100mL
11814163 0.3 μm 15 mL Medium Acid 10% 15mL
11824163 0.3 μm 100 mL Medium Acid 10% 100mL
11834163 0.4 μm 15 mL High Acid 10% 15mL
11844163 0.4 μm 100 mL High Acid 10% 100mL
11894163 0.4 μm 15 mL Low Acid 10% 15mL
11804173 0.4 μm 100 mL Low Acid 10% 100mL
11814173 0.4 μm 1000 mL Low Acid 10% 1000mL
11844203 0.2 μm 15 mL Low Acid 10% 15mL
11854203 0.2 μm 100 mL Low Acid 10% 100mL
11990075 0.5 μm 15 mL Medium Acid 4% 15mL
11900085 0.5 μm 100 mL Medium Acid 4% 100mL
11910085 0.85 μm 15 mL Medium Acid 4% 15mL
11920085 0.85 μm 100 mL Medium Acid 4% 100mL
11804962 0.9 μm 15 mL High Acid 4% 15mL
12493072 0.9 μm 100 mL High Acid 4% 100mL
11814962 4.0 μm 15 mL High Acid 4% 15mL
12433082 4.0 μm 100 mL High Acid 4% 100mL
11824962 5.0 μm 15 mL High Acid 4% 15mL
12453082 5.0 μm 100 mL High Acid 4% 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 30 options available.
30 options
Product Code. 11819922 Supplier Thermo Scientific™ Supplier No. 73003305100250
Control sensitivity, specificity and stability with various acid content available within Thermo Scientific™ Opti-Link™ Carboxylate-Modified Particles.
Control sensitivity, specificity and stability with various acid content available within Thermo Scientific™ Opti-Link™ Carboxylate-Modified Particles. Particles contain carboxylic acid groups for covalent coupling and can be used directly from the bottle in a variety of applications.
Parking area combines surface area and acid content to give you the surface acid distribution that is useful for assay optimization. The surface area per gram of a particle varies inversely with the particle diameter. Acid content refers to the titrated milliequivalents per gram (mEq/g) and is weight-based.
  • For most applications, Opti-Link particles can be used directly from the bottle without any pre-cleaning
  • Proprietary anionic surfactant does not interfere with binding of proteins nor cause proteins to desorb from particle surfaces, nor cause proteins to desorb from particle surfaces
  • Various surface acid concentrations available to help optimize reagent development efforts
  • Opti-Link particles have been used in and are equally successful in molecular biology and research applications
Parking Area Properties
  • Low Acid Content—60–100 Parking Area—Hydrophobic surface, more colloidally stable than plain polystyrene
  • Medium Acid Content—35–59 Parking Area—Hydrophobic and hydrophilic areas; good colloidal stability and good covalent coupling
  • High Acid Content—10–34 Parking Area—Hydrophilic surface; excellent colloidal stability; excellent covalent coupling
Recommended for:
Clinical diagnostics, molecular biology, nephelometric, turbidimetric and slide agglutination assays
Note:
When using Opti-Link particles, ask about our Microparticle Reagent Optimization manual, which outlines our reagent development strategy. This manual includes validated binding, coupling and particle reagent optimization protocols.
TRUSTED_SUSTAINABILITY
Specifications
Additive Proprietary anionic surfactant
Certifications/Compliance Includes Certificate of Analysis
Concentration 10%
Content And Storage Unless otherwise stated, refrigerate (2° to 8°C) product when not in use but do not freeze. Store upright and keep bottle tightly sealed. Mix product with gentle inversion by hand or vortex mixer.
Description Particles that contain carboxylic acid groups for covalent coupling and that can be used in a variety of applications.
For Use With (Application) Covalent Coupling
Material Polystyrene Sulfate
Product Type Specialty Particles
Includes Package Insert Sheet containing Certicate of Analysis
Packaging 15 mL
Particle Density 1.05g/cm³
Preservative 0.05% Na Az
Quantity 15 mL
Sample Evaluation Packs Packaged in 15mL bottles at 10% solids. The evaluation pack consists of three lots from all available PS or CM particles.
Specialty Packaging Opti-Link Carboxylate-Modified Polystyrene particles are available in different concentrations and package sizes upon request.
Surface Acid Loading High Acid
Diameter (Metric) 2 μm