Learn More
Thermo Scientific Chemicals Optical Windows
Product Code. 15437045
Click to view available options
Diameter (Metric):
25.4 mm
50.8 mm
Thickness (Metric):
2 mm
Material:
Cadmium Telluride
Gallium Arsenide
Quartz
Sapphire
Description:
Surface finish: 40/20
Surface finish: 60/40
Unit Size:
Each
Product Code. 15437045
Brand: Thermo Scientific Chemicals 045654.KS
Request Bulk or Custom Format
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Available in different materials and dimensions
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Description Surface finish: 60/40
Material Sapphire
Diameter (Metric) 25.4 mm
Diameter (English) 1 in.
Product Type Optical Window
Thickness (Metric) 2 mm