Learn More
Hellma™ Optical Glass High Precision Macro Absorption Cells With PTFE Stopper
Product Code. 11761744
Click to view available options
Capacity (Metric):
1750 μL
3500 μL
7000 μL
Unit Size:
Each
Pack of 2
Product Code. 11761744
Brand: Hellma™ 1101020
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Specifications
Type Macro Absorption Cell
Capacity (Metric) 3500 μL
Material Optical Glass
Closure Material PTFE
Width (Metric) Interior 9.5mm
Path Length 10 mm
Wavelength Range 320 to 2500 nm
Height (Metric) 46 mm
Width (Metric) 12.5 mm
For Use With (Application) Photometric and Colorimetric Measurements