Description
Achieve maximum reactivity in many diagnostic applications with Thermo Scientific™ Opti-Bind™ Polystyrene Sulfate Particles for use in turbidimetric/nephelometric assays, slide agglutination assays, chemiluminescent assays and in clinical diagnostics and molecular biology applications. Particles can be used direct from the bottle, without any pre-cleaning.
- Available in a wide range of diameters (from 0.1 μM to 2.5 μM)
- No pre-washing
- Tight size distribution—CV<2%
- Optimized colloidal stability
- Shelf-life of 10 years
Recommended for:
Clinical diagnostics, molecular biology, nephelometric, turbidimetric and slide agglutination assays
Note:
When using Opti-Bind particles, ask about our Microparticle Reagent Optimization manual, which outlines our reagent development strategy. This manual includes validated binding, coupling and particle reagent optimization protocols.
Specifications
Specifications
| Certifications/Compliance | Includes Certificate of Analysis |
| Content And Storage | 2°C to 8°C |
| Description | Opti-Bind Polystyrene Sulfate Particles |
| For Use With (Application) | Diagnostic Assays |
| Material | Polystyrene Sulfate |
| Product Type | Specialty Particles |
| Includes | Package Insert and Certificate of Analysis |
| Quantity | 15 mL |
| Sample Evaluation Packs | These evaluation packs allow you to buy small samples from among our standard inventory of different surfaces, colors, sizes and binding capacities at greatly reduced prices. Opt-iBind particles are packaged in 15mL bottles at 10% solids. The evaluation pack consists of three lots from all available Opti-Bind polystyrene particles. |
| Specialty Packaging | Opti-Bind Polystyrene particles are available in different concentrations and package sizes upon request. |
| Show More |