missing translation for 'onlineSavingsMsg'
Learn More

Opianic Acid, TRC

Product Code. 30423642
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30423642 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30423642 Supplier Toronto Research Chemicals Supplier No. TRCO66930050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Opianic acid
CAS 519-05-1
Molecular Formula C10 H10 O5
Quantity 50 mg
Synonym 6-Formyl-2,3-dimethoxybenzoic Acid, 5,6-Dimethoxyphthalaldehydic Acid, 2,3-Dimethoxy-6-formylbenzoic Acid, 5,6-Dimethoxy-2-formylbenzoic Acid, 5,6-Dimethoxyphthalaldehydic Acid, 6-Formyl-2,3-dimethoxybenzoic Acid, NSC 35546, Opianic Acid
InChI Formula InChI=1S/C10H10O5/c1-14-7-4-3-6(5-11)8(10(12)13)9(7)15-2/h3-5H,1-2H3,(H,12,13)
SMILES COc1ccc(C=O)c(C(=O)O)c1OC
IUPAC Name 6-formyl-2,3-dimethoxy-benzoic acid
Molecular Weight (g/mol) 210.18
Formula Weight 210.0528
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.