Learn More
Thermo Scientific™ Open-Top Screw Caps
Product Code. 11567350 Shop All Thermo Scientific Products
Click to view available options
Volume (Metric):
3 mL
3.5 mL
7 mL
10 mL
14 mL
Unit Size:
Pack of 72
Product Code. 11567350
Brand: Thermo Scientific™ TS13216
Thermo Scientific™ Open-Top Screw Caps provide an inert, airtight seal and direct puncture-access to sample with a syringe needle.
Thermo Scientific™ Open-Top Screw Caps provide an inert, airtight seal and direct puncture-access to sample with a syringe needle.
  • Fit into Reacti-Vial™ small reaction vials
  • Used with PTFE/Rubber laminated discs
TRUSTED_SUSTAINABILITY
Specifications
Description TS-13216
Volume (Metric) 7 mL
Type Open Top Screw Cap
Closure Size 15 mm
For Use With (Equipment) 7 mL Screw Cap Septum Vials and Small Reacti-Vials