Learn More
Thermo Scientific™ Open-top Caps without Septum for 8-425 Standard Opening Vials
Product Code. p-4526237
Click to view available options
For Use With (Equipment):
8-425 Standard Opening Screw Vials
8-425 Standard Opening Screw Vials; Flanged for Shimadzu
Unit Size:
Pack of 100
Pack of 1000
Product Code. 12430148
Brand: Thermo Scientific™ C401366
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Thermo Scientific™ National Open-Top Caps without Septum for 8-425 Standard Opening Vials are designed to be used with any 8mm septa.
Thermo Scientific™ National Open-Top Caps without Septum for 8-425 Standard Opening Vials are designed to be used with any 8mm septa.
  • Open top caps for use with any 8mm septa
  • Phenolic caps are suitable for autoclaving and low temperature applications
  • Polypropylene caps are chemically inert and suitable for most chromatography applications
  • Flanged caps are particularly suitable for many Japanese autosamplers
  • Kombo-Kit contains 200 each of black, blue, green, red, and yellow flanged caps
TRUSTED_SUSTAINABILITY
Specifications
Description Phenolic; Black; 8-425
Color Black
Material Phenolic
Type Open Top Screw Cap
Closure Size 8 mm
For Use With (Equipment) 8-425 Standard Opening Screw Vials