Learn More
OHAUS™ Open Ring
OHAUS™ Open Ring designed to securely grip and position laboratory glassware.
Brand:  OHAUS™ 30392344
Additional Details : Weight : 0.50000kg
 View more versions of this product
Product Code. 15782389
  • 39.00€ / Each
Estimated Shipment: 02-07-2024
Description
  • Choose from a variety of specialized LabJaws clamps with adjustable features to provide an exact fit for lab ware such as thermometers, thermocouples, columns, funnels, and round bottom flasks.
  • Versatile multi-purpose LabJaws clamps securely hold glassware and irregularly-shaped objects.
Specifications
Specifications
Aluminum
9 mm
2.280 in.
58.000 mm
Open Ring
Clamp, Specialty, Open Ring, CLS-OPENRAM
6.22 in.
158.00 mm
102 x 102 mm (L x W) Platform