Learn More
OHAUS™ Open Ring
OHAUS™ Open Ring designed to securely grip and position laboratory glassware.
31.00€ - 55.75€
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
15772389
|
OHAUS™
30392343 |
76 x 76 mm (L x W) Platform |
31.00€
Each |
Estimated Shipment: 21-06-2024
Log in to see stock. |
|||||
15782389
|
OHAUS™
30392344 |
102 x 102 mm (L x W) Platform |
39.00€
Each |
Estimated Shipment: 21-06-2024
Log in to see stock. |
|||||
15792389
|
OHAUS™
30392345 |
127 x 127 mm (L x W) Platform |
55.75€
Each |
Estimated Shipment: 21-06-2024
Log in to see stock. |
|||||
Description
- Choose from a variety of specialized LabJaws clamps with adjustable features to provide an exact fit for lab ware such as thermometers, thermocouples, columns, funnels, and round bottom flasks.
- Versatile multi-purpose LabJaws clamps securely hold glassware and irregularly-shaped objects.
Specifications
Aluminum | |
Open Ring |