Learn More
Melford ONPG
Click to view available options
Quantity:
1 g
5 g
25 g
50 g
Unit Size:
1g
25g
50g
5g
Description
Colorimetric and spectrophotometric substrate for detection of β-D-Galactosidase.
Colorimetric and spectrophotometric substrate for detection of β-D-Galactosidase.
Specifications
Specifications
| Quantity | 1 g |