Learn More
Memmert™ ONE™ Oil Baths
Product Code. 10233263 Shop All Memmert Products
Click to view available options
Capacity (Metric):
7 L
10 L
14 L
22 L
29 L
45 L
Unit Size:
Each
Product Code. 10233263
Brand: Memmert™ ONE22
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Protect laboratory and samples, even at high temperatures. Memmert™ ONE™ Oil Baths offers multiple temperature protection through corrosion-resistant stainless steel and precise electronics.
A range of test tube racks, baskets and other accessories is available, please contact Customer Service.
  • Fuzzy supported PID microprocessor controller
  • Digital display
  • 2 x PT100 sensors
  • Calibration facility on controller
  • Audio and visual alarm
  • Over-temperature protection
Specifications
Controller Type Fuzzy-supported PID-microprocessor controller
Capacity (Metric) 22 L
Certifications/Compliance CE, VDE, GS
Display Digital LED
For Use With (Application) For mining, food and beverage, temperature control
Material Stainless Steel
Model ONE 22
Temperature (Metric) Maximum 200°C
Temperature Resolution ±0.1°C
Length (Metric) 578 mm
Length (Metric) Interior 350 mm
Width (Metric) 436 mm
Width (Metric) Interior 290 mm
Height (Metric) 296 mm
Height (Metric) Interior 220 mm
Depth (Metric) Interior 350 mm
Shipping Weight (Metric) 20 kg
Weight (Metric) 16 kg
Voltage 230 V
Wattage 2 w
Frequency 50/60 Hz