Learn More
Ondansetron Hydrochloride Dihydrate, TRC
Specifications
Specifications
| Chemical Name or Material | Ondansetron Hydrochloride Dihydrate |
| CAS | 103639-04-9 |
| Molecular Formula | C18 H19 N3 O . Cl H . 2 H2 O |
| Quantity | 500 mg |
| Synonym | Ondansetron Hydrochloride Dihydrate, 1,2,3,9-Tetrahydro-9-methyl-3-[(2-methyl-1H-imidazol-1-yl)methyl]-4H-carbazol-4-one monohydrochloride dihydrate, Ondansetron hydrochloride dihydrate, Zofran, Zophren |
| InChI Formula | InChI=1S/C18H19N3O.ClH.2H2O/c1-12-19-9-10-21(12)11-13-7-8-16-17(18(13)22)14-5-3-4-6-15(14)20(16)2;;;/h3-6,9-10,13H,7-8,11H2,1-2H3;1H;2*1H2 |
| SMILES | O.O.Cl.Cc1nccn1CC2CCc3c(C2=O)c4ccccc4n3C |
| IUPAC Name | 9-methyl-3-[(2-methylimidazol-1-yl)methyl]-2,3-dihydro-1H-carbazol-4-one;dihydrate;hydrochloride |
| Molecular Weight (g/mol) | 365.85 |
| Formula Weight | 365.1506 |