Learn More
Thermo Scientific™ Oncomine™ NGS Assay Service
Product Code. 18795485 Shop All Thermo Scientific Products
Change view
Click to view available options
1 Sample
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
18795485 1 Sample
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18795485 Supplier Thermo Scientific™ Supplier No. A46080
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Oncomine NGS Assay Service detects genomic variations associated with select cancer hotspots in iPSC samples.
  • Detects genomic variations associated with select cancer hotspots in iPSC samples
  • This service leverages the Ion Torrent Oncomine Comprehensive Assay v3M (A35805) to enable analysis of variants across 161 genes
The Oncomine NGS Assay Service offers:
  • Fast data delivery – turnaround time of four weeks or less
  • Convenient sample prep – just send a frozen cell pellet
  • Detailed results – detection of SNVs, CNVs, gene fusions, and indels
  • Validated data – assay performed in a CLIA-certified clinical laboratory
  • Exceptional support – a dedicated project manager for frequent project communication
TRUSTED_SUSTAINABILITY
Specifications
Quantity 1 sample
Duration 4 Weeks
Product Type NGS Assay Service
Field Research Use Only
Target Audience Cell Line Developers
Species Human