Learn More
Hamilton™ On-Column Injection Syringes with Removable Needles
Click to view available options
Model:
1700 Series
1800 Series
Unit Size:
Each
Description
- On-column injection employs the direct introduction of the liquid sample into the column
- Uses point style 3 syringes
- Point style 3 adapted to the typical septum of the on-column injector
- Needles adjusted to the inner diameter of the column
Specifications
Specifications
| Material | Borosilicate Glass, PTFE |
| Diameter (Metric) Outer | 6.6 mm |
| Product Type | Gastight Syringe |
| Model | 1800 Series |
| Quantity | 1/Ea. |
| Volume (Metric) | 10 μL |
| Needle Point Style | 3 |
| Connection Type | Removable |