Learn More
Fisherbrand™ omniPAGE Blot Insert for omniPAGE Blot MaxiWAVE Vertical Electrophoresis System
Product Code. 12328037 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
For Use With (Equipment):
OmniPAGE Blot MaxiWAVE System
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
12328037 OmniPAGE Blot MaxiWAVE System Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12328037
Estimated Shipment: 09-02-2027
Ensures to maintain even contact between the gel and membrane.
  • Used for Western blotting of proteins
  • Cassettes open architecture ensures the maximum blot area allowing direct transfer of current
Specifications
For Use With (Equipment) OmniPAGE Blot MaxiWAVE System
Type Blot Insert
Includes Three cassettes and 12 fibre pads