Learn More
Diba Omnifit™ Replacement Glass Tubes for EZ and EZ SolventPlus™ Columns
Product Code. 15875304
Encompass_Preferred
Click to view available options
Diameter (Metric) Inner:
6.6 mm
10 mm
15 mm
25 mm
35 mm
50 mm
Length (Metric):
50 mm
100 mm
150 mm
250 mm
330 mm
400 mm
500 mm
Unit Size:
Each
Product Code. 15875304
Brand: Diba 006ERG5033
This item requires a price update prior to being ordered.
Request a Price for this item here.
Mix and match your Omnifit™ Replacement Glass Tubes for EZ and EZ SolventPlus™ Column lengths while using your existing endpieces for ultimate flexibility
Use a different length tube with your existing endpieces to meet your specific requirements.
Specifications
For Use With (Equipment) Omnifit™ EZ and EZ SolventPlus™ Columns
Diameter (Metric) Inner 50 mm
Type Liquid Chromatography Column
Length (Metric) 330 mm
Material Glass
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.