Learn More
Diba Omnifit™ O-Rings for Chromatography Columns
missing translation for 'orderingAttributeHoverText'
Diameter (Metric) Inner:
3 mm
6.6 mm
10 mm
15 mm
25 mm
35 mm
50 mm
Material:
FFKM
FKM
missing translation for 'unitSize'
Pack of 10
Pack of 2
Pack of 6
Specifications
Specifications
| For Use With (Equipment) | Omnifit™ HiT Chromatography Columns |
| Quantity | 6 Pk. |
| Diameter (Metric) Inner | 50 mm |
| Type | Liquid Chromatography Column |
| Length (Metric) | 50 mm |
| Material | FFKM |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.