Learn More
Diba Omnifit™ Frits for EZ, EZ SolventPlus™, BenchMark™, HiT™, and SolventPlus™ Columns
Omnifit™ Frits for EZ, EZ SolventPlus™, BenchMark™, HiT™, and SolventPlus™ Columns feature a range of porosities for your media and chemistry
POA
Specifications
Type | Liquid Chromatography Column |
---|---|
For Use With (Equipment) | Omnifit™ EZ, EZ SolventPlus™, BenchMark™, HiT™, and SolventPlus™ Columns |
Product Code | Brand | Description | Diameter (Metric) Inner | Material | Pore Size | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Diameter (Metric) Inner | Material | Pore Size | Price | Quantity & Availability | ||
15845514
|
Diba
006FR-06-05 |
Chromatography Column Frit, 6.6mm dia, 5 μm, PTFE; 20/pk | 6.6 mm | PTFE | 5 μm |
POA
Pack of 20 |
N/A | ||
This item requires a price update prior to being ordered. Request a Price for this item here. |
|||||||||
15855514
|
Diba
006FR-06-10 |
Chromatography Column Frit, 6.6mm dia, 10 μm, PTFE; 20/pk | 6.6 mm | PTFE | 10 μm |
POA
Pack of 20 |
N/A | ||
This item requires a price update prior to being ordered. Request a Price for this item here. |
|||||||||
15865514
|
Diba
006FR-06-20 |
Chromatography Column Frit, 6.6mm dia, 20 μm, PTFE; 20/pk | 6.6 mm | PTFE | 20 μm |
POA
Pack of 20 |
N/A | ||
This item requires a price update prior to being ordered. Request a Price for this item here. |
|||||||||
15875514
|
Diba
006FR-06-25-PE |
Chromatography Column Frit, 6.6mm dia, 25 μm, Polyethylene; 20/pk | 6.6 mm | Polyethylene | 25 μm |
POA
Pack of 20 |
N/A | ||
This item requires a price update prior to being ordered. Request a Price for this item here. |
|||||||||
15885514
|
Diba
006FR-06-30 |
Chromatography Column Frit, 6.6mm dia, 30 μm, PTFE; 20/pk | 6.6 mm | PTFE | 30 μm |
POA
Pack of 20 |
N/A | ||
This item requires a price update prior to being ordered. Request a Price for this item here. |
|||||||||
15895514
|
Diba
006FR-06-50 |
Chromatography Column Frit, 6.6mm dia, 50 μm, PTFE; 20/pk | 6.6 mm | PTFE | 50 μm |
POA
Pack of 20 |
N/A | ||
This item requires a price update prior to being ordered. Request a Price for this item here. |
|||||||||
15805524
|
Diba
006FR-10-05 |
Chromatography Column Frit, 10mm dia, 5 μm, PTFE; 20/pk | 10 mm | PTFE | 5 μm |
POA
Pack of 20 |
N/A | ||
This item requires a price update prior to being ordered. Request a Price for this item here. |
|||||||||
15815524
|
Diba
006FR-10-10 |
Chromatography Column Frit, 10mm dia, 10 μm, PTFE; 20/pk | 10 mm | PTFE | 10 μm |
POA
Pack of 20 |
N/A | ||
This item requires a price update prior to being ordered. Request a Price for this item here. |
|||||||||
15825524
|
Diba
006FR-10-20 |
Chromatography Column Frit, 10mm dia, 20 μm, PTFE; 20/pk | 10 mm | PTFE | 20 μm |
POA
Pack of 20 |
N/A | ||
This item requires a price update prior to being ordered. Request a Price for this item here. |
|||||||||
15835524
|
Diba
006FR-10-25-PE |
Chromatography Column Frit, 10mm dia, 25 μm, Polyethylene; 20/pk | 10 mm | Polyethylene | 25 μm |
POA
Pack of 20 |
N/A | ||
This item requires a price update prior to being ordered. Request a Price for this item here. |
|||||||||
15845524
|
Diba
006FR-10-30 |
Chromatography Column Frit, 10mm dia, 30 μm, PTFE; 20/pk | 10 mm | PTFE | 30 μm |
POA
Pack of 20 |
N/A | ||
This item requires a price update prior to being ordered. Request a Price for this item here. |
|||||||||
15855524
|
Diba
006FR-10-50 |
Chromatography Column Frit, 10mm dia, 50 μm, PTFE; 20/pk | 10 mm | PTFE | 50 μm |
POA
Pack of 20 |
N/A | ||
This item requires a price update prior to being ordered. Request a Price for this item here. |
|||||||||
15885524
|
Diba
006FR-15-20 |
Chromatography Column Frit, 15mm dia, 20 μm, PTFE; 20/pk | 15 mm | PTFE | 20 μm |
POA
Pack of 20 |
N/A | ||
This item requires a price update prior to being ordered. Request a Price for this item here. |
|||||||||
15805534
|
Diba
006FR-15-30 |
Chromatography Column Frit, 15mm dia, 30 μm, PTFE; 20/pk | 15 mm | PTFE | 30 μm |
POA
Pack of 20 |
N/A | ||
This item requires a price update prior to being ordered. Request a Price for this item here. |
|||||||||
15815534
|
Diba
006FR-15-50 |
Chromatography Column Frit, 15mm dia, 50 μm, PTFE; 20/pk | 15 mm | PTFE | 50 μm |
POA
Pack of 20 |
N/A | ||
This item requires a price update prior to being ordered. Request a Price for this item here. |
|||||||||
15825534
|
Diba
006FR-25-05 |
Chromatography Column Frit, 25mm dia, 5 μm, PTFE; 20/pk | 25 mm | PTFE | 5 μm |
POA
Pack of 20 |
N/A | ||
This item requires a price update prior to being ordered. Request a Price for this item here. |
|||||||||
15835534
|
Diba
006FR-25-10 |
Chromatography Column Frit, 25mm dia, 10 μm, PTFE; 20/pk | 25 mm | PTFE | 10 μm |
POA
Pack of 20 |
N/A | ||
This item requires a price update prior to being ordered. Request a Price for this item here. |
|||||||||
15845534
|
Diba
006FR-25-20 |
Chromatography Column Frit, 25mm dia, 20 μm, PTFE; 20/pk | 25 mm | PTFE | 20 μm |
POA
Pack of 20 |
N/A | ||
This item requires a price update prior to being ordered. Request a Price for this item here. |
|||||||||
15855534
|
Diba
006FR-25-25-PE |
Chromatography Column Frit, 25mm dia, 25 μm, Polyethylene; 20/pk | 25 mm | Polyethylene | 25 μm |
POA
Pack of 20 |
N/A | ||
This item requires a price update prior to being ordered. Request a Price for this item here. |
|||||||||
15865534
|
Diba
006FR-25-30 |
Chromatography Column Frit, 25mm dia, 30 μm, PTFE; 20/pk | 25 mm | PTFE | 30 μm |
POA
Pack of 20 |
N/A | ||
This item requires a price update prior to being ordered. Request a Price for this item here. |
|||||||||
15875534
|
Diba
006FR-25-50 |
Chromatography Column Frit, 25mm dia, 50 μm, PTFE; 20/pk | 25 mm | PTFE | 50 μm |
POA
Pack of 20 |
N/A | ||
This item requires a price update prior to being ordered. Request a Price for this item here. |
|||||||||
15805544
|
Diba
006FR-35-20 |
Chromatography Column Frit, 35mm dia, 20 μm, PTFE; 20/pk | 35 mm | PTFE | 20 μm |
POA
Pack of 20 |
N/A | ||
This item requires a price update prior to being ordered. Request a Price for this item here. |
|||||||||
15825544
|
Diba
006FR-35-30 |
Chromatography Column Frit, 35mm dia, 30 μm, PTFE; 20/pk | 35 mm | PTFE | 30 μm |
POA
Pack of 20 |
N/A | ||
This item requires a price update prior to being ordered. Request a Price for this item here. |
|||||||||
15835544
|
Diba
006FR-35-50 |
Chromatography Column Frit, 35mm dia, 50 μm, PTFE; 20/pk | 35 mm | PTFE | 50 μm |
POA
Pack of 20 |
N/A | ||
This item requires a price update prior to being ordered. Request a Price for this item here. |
|||||||||
15865544
|
Diba
006FR-50-20 |
Chromatography Column Frit, 50mm dia, 20 μm, PTFE; 20/pk | 50 mm | PTFE | 20 μm |
POA
Pack of 20 |
N/A | ||
This item requires a price update prior to being ordered. Request a Price for this item here. |
|||||||||
15885544
|
Diba
006FR-50-30 |
Chromatography Column Frit, 50mm dia, 30 μm, PTFE; 20/pk | 50 mm | PTFE | 30 μm |
POA
Pack of 20 |
N/A | ||
This item requires a price update prior to being ordered. Request a Price for this item here. |
|||||||||
15895544
|
Diba
006FR-50-50 |
Chromatography Column Frit, 50mm dia, 50 μm, PTFE; 20/pk | 50 mm | PTFE | 50 μm |
POA
Pack of 20 |
N/A | ||
This item requires a price update prior to being ordered. Request a Price for this item here. |
|||||||||
Specifications
Liquid Chromatography Column | |
Omnifit™ EZ, EZ SolventPlus™, BenchMark™, HiT™, and SolventPlus™ Columns |