Learn More
Cytiva Omega Membrane Discs
Product Code. 17194811
Change view
Click to view available options
MWCO:
1 kDa
10 kDa
100 kDa
30 kDa
5 kDa
Diameter (Metric) Membrane:
43 mm
62 mm
76 mm
Unit Size:
Pack of 12
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. MWCO Diameter (Metric) Membrane Unit Size
17194811 30 kDa 62 mm Pack of 12
17154811 10 kDa 43 mm Pack of 12
17184811 30 kDa 43 mm Pack of 12
17174811 10 kDa 76 mm Pack of 12
17134811 5 kDa 43 mm Pack of 12
17104821 30 kDa 76 mm Pack of 12
17144811 5 kDa 76 mm Pack of 12
17114811 1 kDa 43 mm Pack of 12
17124811 1 kDa 76 mm Pack of 12
17164811 10 kDa 62 mm Pack of 12
17114821 100 kDa 43 mm Pack of 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Product Code. 17194811 Supplier Cytiva Supplier No. OM030062
  • ONLINE EXCLUSIVE PRICE
    435.00€
    407.31€ / Pack of 12
    Save 27.69€
    6% Off
 Restricted Item
Estimated Shipment: 28-12-2026
Omega Membrane Discs with high performance and stability for users of conventional, non-disposable stirred cells.
  • Polyethersulfone membrane provides higher flow rates and lower protein binding than competitive regenerated cellulose
  • Lower processing times and the highest possible recoveries
  • Discs can be washed and reused
  • Replacement membranes for other stirred cell systems concentration, fractionation, purification, buffer exchange, or desalting of protein, cell broths, and other biomolecules
  • For research use only
TRUSTED_SUSTAINABILITY
Specifications
Material (Membrane) PES
MWCO 30 kDa
Sterility Non-sterile
No. per Pack 12/Pk.
Diameter (Metric) Membrane 62 mm