missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ OLFM1 Recombinant Protein

Brand:  Abnova™ H00010439-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16165092

  • 359.00€ / 10µg

Estimated Shipment: 22-08-2022
to see stock.
Add to basket

Description

Description

ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array

Specifications

Specifications

AAH00189
Solution
10439
OLFM1 (Human) Recombinant Protein (P01)
In vitro wheat germ expression system
12.5% SDS-PAGE Stained with Coomassie Blue.
Wheat Germ (in vitro)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
OLFM1
Human
Yes
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
44.33
8
Glutathione Sepharose 4 Fast Flow
10 ug
MPGRWRWQRDMHPARKLLSLLFLILMGTELTQNKRENKAEKMGGPESERKTTGEKTLNELPLFCLEAHAGSLALPRMCSPNPNPAVGLCRPAYPQSPSPGAAQTISQSLLERFCMASRREVFLAPGRPGGGWWLCTVAGQMHSFMCTHTHTHAHTGEQIPAEKSQPGPD
AMY/NOE1/NOELIN1/OlfA
OLFM1
Wheat Germ (in vitro)
GST
Videos
SDS
Documents
Certificates
Special Offers

Special Offers