missing translation for 'onlineSavingsMsg'
Learn More

Oleyl Oleate, TRC

Product Code. 30412873
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30412873 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30412873 Supplier Toronto Research Chemicals Supplier No. TRCO5323005G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Oley Oleate
CAS 3687-45-4
Molecular Formula C36 H68 O2
Quantity 5 g
Synonym 9-Octadecenoic acid (9Z)-','(9Z)-9-octadecen-1-yl ester (ACI), 9-Octadecen-1-ol','oleate','(Z)- (8CI), 9-Octadecenoic acid (9Z)-','(9Z)-9-octadecenyl ester (9CI), 9-Octadecenoic acid (Z)-','9-octadecenyl ester','(Z)- (ZCI), Oleic acid','(Z)-9-octadecenyl ester (8CI), Oleic acid','9-octadecenyl ester (6CI), Oleic acid','cis-9-octadecenyl ester (7CI), Cetiol, Cosmol liquid, Crodamol OO-V, Exceparl OL-OL, Oleic acid oleyl ester, Oleyl oleate, Schercemol OLO, Oleic Acid','9-octadecenyl Ester
InChI Formula InChI=1S/C36H68O2/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33-35-38-36(37)34-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h17-20H,3-16,21-35H2,1-2H3/b19-17-,20-18-
SMILES CCCCCCCC\C=C/CCCCCCCCOC(=O)CCCCCCC\C=C/CCCCCCCC
IUPAC Name [(Z)-octadec-9-enyl] (Z)-octadec-9-enoate
Molecular Weight (g/mol) 532.92
Formula Weight 532.5219
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.