missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Oleyl Oleate, TRC
Specifications
Specifications
| Chemical Name or Material | Oley Oleate |
| CAS | 3687-45-4 |
| Molecular Formula | C36 H68 O2 |
| Quantity | 500 mg |
| Synonym | 9-Octadecenoic acid (9Z)-','(9Z)-9-octadecen-1-yl ester (ACI), 9-Octadecen-1-ol','oleate','(Z)- (8CI), 9-Octadecenoic acid (9Z)-','(9Z)-9-octadecenyl ester (9CI), 9-Octadecenoic acid (Z)-','9-octadecenyl ester','(Z)- (ZCI), Oleic acid','(Z)-9-octadecenyl ester (8CI), Oleic acid','9-octadecenyl ester (6CI), Oleic acid','cis-9-octadecenyl ester (7CI), Cetiol, Cosmol liquid, Crodamol OO-V, Exceparl OL-OL, Oleic acid oleyl ester, Oleyl oleate, Schercemol OLO, Oleic Acid','9-octadecenyl Ester |
| InChI Formula | InChI=1S/C36H68O2/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33-35-38-36(37)34-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h17-20H,3-16,21-35H2,1-2H3/b19-17-,20-18- |
| SMILES | CCCCCCCC\C=C/CCCCCCCCOC(=O)CCCCCCC\C=C/CCCCCCCC |
| IUPAC Name | [(Z)-octadec-9-enyl] (Z)-octadec-9-enoate |
| Molecular Weight (g/mol) | 532.92 |
| Formula Weight | 532.5219 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?