missing translation for 'onlineSavingsMsg'
Learn More

Olanzapine-D3 1.0 mg/ml in Acetonitrile, LoGiCal

Product Code. 30460234
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 30460234

Brand: LoGiCal LGCAMP0329.8111

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Olanzapine-methyl-D3
CAS 786686-79-1
Solvent or Matrix Acetonitrile
Molecular Formula C17 2H3 H17 N4 S
Quantity 1 mL
Synonym 2-Methyl-4-(4-methyl-d3-1-piperazinyl)-10H-thieno[2,3-b][1,5]benzodiazepine, Integrol-d3, LY 170053-d3, Lanzac-d3, Oleanz-d3, Oliza-d3, Oltal-d3, Zyprexa-d3, 2-Methyl-4-[4-(methyl-d3)-1-piperazinyl]-10H-thieno[2,3-b][1,5]benzodiazepine (9CI), Olanzapine-D3
InChI Formula InChI=1S/C17H20N4S/c1-12-11-13-16(21-9-7-20(2)8-10-21)18-14-5-3-4-6-15(14)19-17(13)22-12/h3-6,11,19H,7-10H2,1-2H3/i2D3
SMILES [2H]C([2H])([2H])N1CCN(CC1)C2=Nc3ccccc3Nc4sc(C)cc24
IUPAC Name 2-methyl-4-[4-(trideuteriomethyl)piperazin-1-yl]-10H-thieno[2,3-b][1,5]benzodiazepine
Molecular Weight (g/mol) 315.45
Formula Weight 315.1597 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.