missing translation for 'onlineSavingsMsg'
Learn More

DuraLab OIML M1 Stainless Steel Calibration Weight

Product Code. 30280305
Click to view available options
Weight (Metric):
200 g
500 g
2000 g
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 30280305

Brand: DuraLab DRL0053

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The OIML-certified DuraLab M1 Calibration Weight is designed for use with the DuraLab Balance range. It is ideal for laboratories, educational institutions, and industrial environments where precision is paramount.

  • Made from stainless steel for durability and corrosion resistance
  • Supports accurate calibration of laboratory balances
  • Meets OIML M1 tolerance class standards
  • Supplied in a protective plastic container
  • Helps maintain measurement accuracy in laboratory, industrial, and educational settings
  • Recommended for calibrating DuraLab balances
  • Balances are not supplied with calibration weights as standard

Specifications

Product Type Calibration Weight
Weight (English) 7.05 oz.
Material Stainless Steel
Weight (Metric) 200 g
Tolerance ±100 mg
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.