Learn More
Sartorius™ OIML Class M1 Proof Line Knob Weight without Certificate
Product Code. 11303424 Shop All Sartorius Products
Click to view available options
Mass:
1 g
2 g
5 g
10 g
20 g
50 g
200 g
500 g
1 kg
2 kg
Unit Size:
Each
Product Code. 11303424
Brand: Sartorius™ YCW555AC00
Only null left
Add to basket
Add to basket
Polished non-magnetizable calibration weights. Sartorius™ OIML Class M1 Proof Line Knob Weight without Certificate is constructed of stainless steel and supplied in a plastic case.
  • Non-magnetizable
  • Supplied in a high-quality plastic case (unless otherwise specified) suitable for use in cleanrooms
  • Stainless steel construction with a density of 7,850 kg/m3
  • Complies with OIML R111-1 International Recommentation
TRUSTED_SUSTAINABILITY
Specifications
Product Type Proof Line Weight
Classification OIML Class M1
Material Stainless Steel
Density 7,850kg/m3
Mass 500 g