Learn More
Kern Sohn OIML Class M1 ECO-Block Test Weight
Description
- ECO rectangular shape
- Lacquered cast iron
Specifications
Specifications
| Classification | M1 |
| Material | Cast Iron |
| Weight (Metric) | 10 kg |
| Tolerance | ± 500 mg |