Learn More
Kern Sohn OIML Class M1 ECO-Block Test Weight
Product Code. 17342443
Encompass_Preferred
Change view
Click to view available options
Weight (Metric):
5 kg
10 kg
20 kg
50 kg
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Weight (Metric) unitSize
17342443 5 kg Each
17352443 10 kg Each
17362443 20 kg Each
17372443 50 kg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 17342443 Supplier Kern Sohn Supplier No. T34676A
Single test weight constructed of cast iron.
  • ECO rectangular shape
  • Lacquered cast iron
Specifications
Classification M1
Material Cast Iron
Weight (Metric) 5 kg
Tolerance ± 250 mg