Learn More
Rogo Sampaic OIL BATH ROUND 2,5L
Brand:  Rogo Sampaic JDB502
OIL BATH ROUND 2,5L
Product Code. 11892702
  • ONLINE EXCLUSIVE PRICE
    1255.00€
    649.76€ / Each
    Save 605.24€
    48% Off
Only null left
Estimated Shipment: 16-03-2026