Learn More
N-Acetyl Amonafide, TRC
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Specifications
Specifications
| Chemical Name or Material | N-Acetyl Amonafide |
| CAS | 69409-02-5 |
| Molecular Formula | C18H19N3O3 |
| Quantity | 5 mg |
| Synonym | N-[2-[2-(Dimethylamino)ethyl]-2,3-dihydro-1,3-dioxo-1H-benz[de]isoquinolin-5-yl]acetamide, MADE-FA 4512, N-Acetylamonafide |
| InChI Formula | InChI=1S/C18H19N3O3/c1-11(22)19-13-9-12-5-4-6-14-16(12)15(10-13)18(24)21(17(14)23)8-7-20(2)3/h4-6,9-10H,7-8H2,1-3H3,(H,19,22) |
| SMILES | O=C(C1=C2C(C=CC=C23)=CC(NC(C)=O)=C1)N(CCN(C)C)C3=O |
| IUPAC Name | N-[2-[2-(dimethylamino)ethyl]-1,3-dioxobenzo[de]isoquinolin-5-yl]acetamide |
| Molecular Weight (g/mol) | 325.36 |