Learn More
OHAUS™ Sample Pan for MB Series Moisture Analyzers
Product Code. 11865390 Shop All Ohaus Products
Change view
Click to view available options
Type:
Disposable Sample Pan
Reusable Sample Pan
Quantity:
3/Pk.
80/Pk.
Unit Size:
Pack of 3
Pack of 80
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Quantity unitSize
11865390 Reusable Sample Pan 3/Pk. Pack of 3
11804242 Disposable Sample Pan 80/Pk. Pack of 80
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11865390 Supplier OHAUS™ Supplier No. 80850088
Only null left
Add to basket
Add to basket
Sample Pans for OHAUS™ MB series moisture analyzers.
  • Shape: round
  • Choose disposable or reusable
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) US: MB35,MB45,MB27,MB90,MB120 Moisture Analyzers
EU: MB27, MB25, MB90-GB, MB120-GB, MB23, MB120-EU
Quantity 3/Pk.
Type Reusable Sample Pan