missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Octyl Phenol Ethoxylate, BAKER ANALYZED™ Reagent, J.T. Baker™
Shop All J.T.Baker Products
Click to view available options
Quantity:
120 mL
Packaging:
Narrow Mouth Amber Glass Bottle
Unit Size:
120mL
Specifications
Specifications
| Flash Point | 251.00°C |
| Packaging | Narrow Mouth Amber Glass Bottle |
| Quantity | 120 mL |
| Specific Gravity | 1L = 1.07kg |
| Grade | BAKER ANALYZED™ Reagent |
| Chemical Name or Material | Octyl Phenol Ethoxylate |
Safety and Handling
Product Identifier
- Octyl phenol ethoxylate
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- P264-Wash hands thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P321-Specific treatment (see on this label).
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P362+P364-Take off contaminated clothing and wash it before reuse.
- MIXTURE LIST-Contains : Octyl phenol ethoxylate
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction