missing translation for 'onlineSavingsMsg'
Learn More

Octamethylcyclotetrasiloxane, TRC

Product Code. 30135981
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135981 25 g 25g
30040592 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30135981 Supplier Toronto Research Chemicals Supplier No. TRCO23714525G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 556-67-2
Formula Weight 296.0752
InChI Formula InChI=1S/C8H24O4Si4/c1-13(2)9-14(3,4)11-16(7,8)12-15(5,6)10-13/h1-8H3
IUPAC Name 2,2,4,4,6,6,8,8-octamethyl-1,3,5,7,2,4,6,8-tetraoxatetrasilocane
Molecular Formula C8 H24 O4 Si4
Molecular Weight (g/mol) 296.62
Percent Purity >95%
SMILES C[Si]1(C)O[Si](C)(C)O[Si](C)(C)O[Si](C)(C)O1
Synonym Cyclotetrasiloxane, octamethyl- (8CI,9CI), 2,2,4,4,6,6,8,8-Octamethylcyclotetrasiloxane, 2,2,4,4,6,6,8,8-Octamethyl-1,3,5,7,2,4,6,8-tetraoxatetrasilocane, Abil K 4, Additive 8030, Cyclic dimethylsiloxane tetramer, D 4, DC 244, DC 344, DC 5258, DWQ 110, DWQ 120, Dabco DC 5258, Dow Corning 244, Dow Corning 244 Fluid, Dow Corning 344, KF 994, LS 8620, Mirasil CM 4, NSC 345674, NUC Silicone VS 7207, Octamethylcyclotetrasiloxane, Octamethylcyclotetrasiloxanes, PMX 0110, SF 1173, SH 244, SH 244 Fluid, SH 30, SH 344, Silbione 70045V2, Silbione V 2, TSF 404, Tetracyclomethicone, UC 7207, Union Carbide 7207, VS 7207, Volasil 244, XF 3803, Y 7175, YF 393, ZBH 270
Chemical Name or Material Octamethylcyclotetrasiloxane
Grade GC
Show More Show Less

Specifications

Quantity 25 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.