Learn More
Octamethyl-1,7-tetrasiloxanediol, TRC
Product Code. 30414331
Change view
Click to view available options
Quantity:
25 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30414331 100 mg
30411249 250 mg
30412134 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30414331 Supplier Toronto Research Chemicals Supplier No. TRCO860130100MG
 Restricted Item
Estimated Shipment: 12-11-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 3081-07-0
Molecular Formula C8H26O5Si4
Molecular Weight (g/mol) 314.63
Synonym 1,1,3,3,5,5,7,7-Octamethyl-1,7-tetrasiloxanediol, 1,7-Dihydroxyoctamethyltetrasiloxane
IUPAC Name hydroxy-[[[hydroxy(dimethyl)silyl]oxy-dimethylsilyl]oxy-dimethylsilyl]oxy-dimethylsilane
SMILES C[Si](C)(O)O[Si](C)(C)O[Si](C)(C)O[Si](C)(C)O
Specifications
Quantity 100 mg
InChI Formula InChI=1S/C8H26O5Si4/c1-14(2,9)11-16(5,6)13-17(7,8)12-15(3,4)10/h9-10H,1-8H3
Formula Weight 314.0857
Chemical Name or Material Octamethyl-1,7-tetrasiloxanediol