Learn More
Macherey-Nagel™ Octadecyl Phase Polygosil™
Click to view available options
Quantity:
10 g
100 g
Unit Size:
100g
10g
Specifications
Specifications
| Particle Size | 7 μm |
| Pore Size | 100 Å |
| Technique | Liquid Chromatography |
| Quantity | 10 g |
| Type | Polygosil C18 Phases |