Learn More
Octadecanoyl-L-carnitine-d3 HCl (N-methyl-d3), CDN
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
Specifications
Specifications
| Chemical Name or Material | Stearoyl-L-carnitine-d3 Hydrochloride |
| CAS | 2245711-27-5 |
| Molecular Formula | C25 D3 H47 N O4 . Cl |
| Quantity | 0.01 g |
| Synonym | 1-Propanaminium, 3-carboxy-N,N-dimethyl-N-(methyl-d3)-2-[(1-oxooctadecyl)oxy]-, chloride (1:1), (2 R)- (ACI), Octadecanoyl-L-carnitine-d3 HCl (N-methyl-d3), O-Octadecanoyl-L-carnitine-d3 chloride, Stearoyl-L-carnitine-d3 chloride, Stearoyl-L-carnitine-D3 (N-methyl-D3) chloride, (2 R)-3-Carboxy-N,N-dimethyl-N-(methyl-d3)-2-[(1-oxooctadecyl)oxy]-1-propanaminium chloride |
| InChI Formula | InChI=1 S/C25H49NO4.ClH/c1-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-25(29)30-23(21-24(27)28)22-26(2,3)4;/h23H,5-22H2,1-4H3;1 H/t23-;/m1./s1/i2D3; |
| SMILES | [Cl-].[2 H]C([2 H])([2 H])[N+](C)(C)C[C@@H](CC(=O)O)OC(=O)CCCCCCCCCCCCCCCCC |
| IUPAC Name | [(2 R)-3-carboxy-2-octadecanoyloxypropyl]-dimethyl-(trideuteriomethyl)azanium;chloride |
| Molecular Weight (g/mol) | 467.14 |
| Formula Weight | 466.3617 g/mol |
| Show More |