missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Octadecanoic-17,17,18,18,18-d5 Acid, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | Octadecanoic-17,17,18,18,18-D5 Acid |
| CAS | 211443-83-3 |
| Molecular Formula | C18H31D5O2 |
| Quantity | 0.1 g |
| Synonym | Octadecanoic-17,17,18,18,18-d5 Acid, Octadecanoic-17,17,18,18,18-d5 acid (9 CI), Stearic-17,17,18,18,18-d5 acid, 17,17,18,18,18-Pentadeuterostearic acid, [17,17,18,18,18-d5]-Stearic acid, Stearic acid-17,17,18,18,18-d5, Octadecanoic-17,17,18,18,18-D5 acid |
| InChI Formula | InChI=1 S/C18H36O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20/h2-17H2,1H3,(H,19,20)/i1D3,2D2 |
| SMILES | OC(CCCCCCCCCCCCCCCC([2 H])([2 H])C([2 H])([2 H])[2 H])=O |
| IUPAC Name | octadecanoic-17,17,18,18,18-d5 acid |
| Molecular Weight (g/mol) | 289.51 |
| Formula Weight | 289.3 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction