Learn More
Octadecanoic-16,16,17,17,18,18,18-d7 Acid, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Specifications
Specifications
| Chemical Name or Material | Octadecanoic-16,16,17,17,18,18,18-D7 Acid |
| CAS | 951209-59-9 |
| Molecular Formula | C18 D7 H29 O2 |
| Quantity | 0.1 g |
| Synonym | Octadecanoic-16,16,17,17,18,18,18-d7 acid (ACI), 16,16,17,17,18,18,18-d7-Stearic acid, 16,16,17,17,18,18,18-Heptadeuterostearic acid, [16,16,17,17,18,18,18-2H7]Stearic acid, Stearic acid-16,16,17,17,18,18,18-d7 |
| InChI Formula | InChI=1 S/C18H36O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20/h2-17H2,1H3,(H,19,20)/i1D3,2D2,3D2 |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])CCCCCCCCCCCCCCC(=O)O |
| IUPAC Name | 16,16,17,17,18,18,18-heptadeuteriooctadecanoic acid |
| Molecular Weight (g/mol) | 291.5204 |
| Formula Weight | 291.3155 g/mol |
| Show More |