missing translation for 'onlineSavingsMsg'
Learn More

Octadecanamide, TRC

Product Code. 30422207
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30422207 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30422207 Supplier Toronto Research Chemicals Supplier No. TRCO2350031G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Octadecanamide
CAS 124-26-5
Molecular Formula C18H37NO
Quantity 1 g
Synonym Stearamide (6CI,7CI,8CI), Adogen 42, Advawax 290, Alflow S 10, Alflow S 100, Amide AP, Amide S (binder), Amide T, Armid 18, Armoslip 18LF, Atmer SA 1750, Blaunon SD 50, Chamide S, Chamide SSA, Crodamide S, Crodamide SR, Daiwax SA 200, EBF, Fatty Amide T, Finawax S, Finawax S 70, Finawax S 90, G 270, Hidorin B 934, Hidorin F 792, Hidorin L 271, Hidorin M 7, Hymicron G 270, Hymicron L 271, Impoe HE 88, Kao Fatty Amide S, Kawaslip VL, Kemamide S, L 271, Loxamid S, Lutamide S, Lutamide SR, N 327, NSC 66462, Neutron 2, Neutron 2S, Octadecamide, Octadecaneamide, Octadecylamide, Selosol 2M, Stearic acid amide, Stearic amide, Stearoylamide, Stearoylamine, Stearylamide, T 22, T 22 (amide), Uniwax 1750, Stearamide, Stearlyamide, Stearic Acid Amide, Octadecylamide
InChI Formula InChI=1S/C18H37NO/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20/h2-17H2,1H3,(H2,19,20)
SMILES CCCCCCCCCCCCCCCCCC(=O)N
IUPAC Name octadecanamide
Molecular Weight (g/mol) 283.49
Formula Weight 283.2875
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.