Learn More
Ochromycinone, TRC
Specifications
Specifications
| Chemical Name or Material | Ochromycinone |
| CAS | 28882-53-3 |
| Molecular Formula | C19 H14 O4 |
| Quantity | 0.5 mg |
| InChI Formula | InChI=1S/C19H14O4/c1-9-7-10-5-6-12-17(15(10)14(21)8-9)19(23)11-3-2-4-13(20)16(11)18(12)22/h2-6,9,20H,7-8H2,1H3/t9-/m0/s1 |
| SMILES | C[C@@H]1CC(=O)c2c(C1)ccc3C(=O)c4c(O)cccc4C(=O)c23 |
| IUPAC Name | (3S)-8-hydroxy-3-methyl-3,4-dihydro-2H-benzo[a]anthracene-1,7,12-trione |
| Molecular Weight (g/mol) | 306.31 |
| Formula Weight | 306.0892 |