Learn More
Motic™ Objectives, Stereo for SMZ-161/171
Product Code. 15465442
Click to view available options
Magnification Power:
0.3X
0.63X
0.75X
1.5X
Description:
WD: 128.6mm
WD: 142.7mm
WD: 301mm
WD: 56.3mm
Unit Size:
Each
Product Code. 15465442
Brand: Motic™ 1101001703661
Suitable stereo objectives for SMZ-161 and SMZ-171 Microscopes. Motic™ Objectives, Stereo for SMZ-161/171 are precision-crafted for superior quality and durability.
  • Suitable for SMZ-161 and SMZ-171 Microscopes.
Specifications
Type Stereo
Description WD: 142.7mm
For Use With (Equipment) SMZ-161/171 Microscopes
Magnification Power 0.63X