Learn More
Zeiss™ Objective Plan Achromat
Click to view available options
Type:
Objective Plan-Achromat 10X/0.25 Ph1
Objective Plan-Achromat 20X/0.40
Objective Plan-Achromat 4X/0.10
Magnification Power:
10X
20X
4x
Description:
WD: 0.9mm
WD: 12mm
WD: 4.4mm
Unit Size:
Each
Description
- Objective Plan-Achromat 20x/0.40 for Primo (WD: 0.90mm)
- Objective Plan-Achromat 10x/0.25 Ph1 for Primo (WD: 4.4mm)
- Objective Plan-Achromat 4x/0.10 for Primo (WD: 12.0mm)
Specifications
Specifications
| Type | Objective Plan-Achromat 20X/0.40 |
| Description | WD: 0.9mm |
| For Use With (Equipment) | Primo |
| Magnification Power | 20X |
| Quantity | 1 Ea. |