Learn More
Zeiss™ Objective Nosepiece
Click to view available options
Type:
Objective Nosepiece S/doc
Unit Size:
Each
Description
- Objective nosepiece S/doc
- 3 fold, 6x cod
- For stereo and axial mode using 3 objectives on SteREO Discovery
- Mount at focus drive with Profile S column as an alternative of 435402 Carrier S dia.: 76mm or 435116 Co-observer equipment
Specifications
Specifications
| Type | Objective Nosepiece S/doc |
| For Use With (Equipment) | Stero Discovery Microscope |
| Magnification Power | 6X |
| Quantity | 1 Ea. |