Learn More
Zeiss™ Objective LD A-Plan
Designed for routine research. A-Plan Objectives are ideal for a variety of applications.
Brand:  Zeiss™ 4212319910
Additional Details : Weight : 0.50000kg
 View more versions of this product
Product Code. 14801251
  • 834.00€ / Each
Estimated Shipment: 20-06-2024
Description
  • This class of objectives provides an attractive alternative to “Achroplan” objectives, both in routine and research applications
  • “A-Plan” objectives provide rich contrast and can be used with eyepieces with a field of view up to 23mm
  • Allow contrasting methods ranging from brightfield to phase contrast
  • For excitation wavelength within the visible spectrum they can be used for epifluorescence work
Specifications
Specifications
Objective LD A-Plan
4.3 mm
20X